Anti-AIF1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029412EA 590 EUR
USBI029412
Anti-AIF1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.1 ?g/ml; Immunochemistry: 1:1000

Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR