Anti-BMPR1B Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029428EA 549 EUR
USBI029428
Anti-BMPR1B Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: BMPR1B (bone morphogenetic protein receptor, type IB)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR