Anti-DRAM1 Mouse Monoclonal Antibody [clone: 13A174]

Lieferant: United States Biological
USBI029455EA 550 EUR
USBI029455
Anti-DRAM1 Mouse Monoclonal Antibody [clone: 13A174]
Antikörper
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:400

Type: Primary
Antigen: DRAM1 (DNA-damage regulated autophagy modulator 1)
Clonality: Monoclonal
Clone: 13A174
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR