Anti-DRAM2 Mouse Monoclonal Antibody [clone: 13A175]

Lieferant: United States Biological
USBI029456EA 550 EUR
USBI029456
Anti-DRAM2 Mouse Monoclonal Antibody [clone: 13A175]
Antikörper
Recommended dilution: Western Blot:1 ?g/ml

Type: Primary
Antigen: DRAM2 (DNA-damage regulated autophagy modulator 2)
Clonality: Monoclonal
Clone: 13A175
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR