Anti-GSK3A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A190]

Lieferant: United States Biological
USBI029488EA 488 EUR
USBI029488
Anti-GSK3A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A190]
Antikörper
Type: Primary
Antigen: GSK3A (glycogen synthase kinase 3 alpha)
Clonality: Monoclonal
Clone: 13A190
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR