Anti-Glutathione S Transferase Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029489EA 519 EUR
USBI029489
Anti-Glutathione S Transferase Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0. 05 ?g/ml

Type: Tag
Antigen: Glutathione S Transferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR