Anti-Histone H4, acetylated Rabbit Monoclonal Antibody [clone: 13A196]

Lieferant: United States Biological
USBI029504EA 640 EUR
USBI029504
Anti-Histone H4, acetylated Rabbit Monoclonal Antibody [clone: 13A196]
Antikörper
Type: Primary
Antigen: Histone H4, acetylated
Clonality: Monoclonal
Clone: 13A196
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR