Anti-Histone H4, Dimethyl Symmetric Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029505EA 640 EUR
USBI029505
Anti-Histone H4, Dimethyl Symmetric Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Histone H4, Dimethyl Symmetric
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR