Anti-HSNF2H Mouse Monoclonal Antibody [clone: 13A199]

Lieferant: United States Biological
USBI029509EA 561 EUR
USBI029509
Anti-HSNF2H Mouse Monoclonal Antibody [clone: 13A199]
Antikörper
Recommended dilution: Western Blot: 0.5 ?g/ml; Immunochemistry: 1:500

Type: Primary
Antigen: HSNF2H
Clonality: Monoclonal
Clone: 13A199
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR