Anti-IGF2BP3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029512EA 674 EUR
USBI029512
Anti-IGF2BP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGF2BP3 (insulin-like growth factor 2 mRNA binding protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR