Anti-INF2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029515EA 716 EUR
USBI029515
Anti-INF2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0. 05 ?g/ml; Immunochemistry: 1:500

Type: Primary
Antigen: INF2 (inverted formin, FH2 and WH2 domain containing)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR