Anti-NES Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13A210]

Lieferant: United States Biological
USBI029552EA 499 EUR
USBI029552
Anti-NES Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13A210]
Antikörper
Do not freeze!

Type: Primary
Antigen: NES (nestin)
Clonality: Monoclonal
Clone: 13A210
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR