Anti-PRDM14 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029588EA 713 EUR
USBI029588
Anti-PRDM14 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PRDM14 (PR domain containing 14)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR