Anti-RAB25 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029594EA 545 EUR
USBI029594
Anti-RAB25 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0. 05 ?g/ml; Immunochemistry: 1:600

Type: Primary
Antigen: RAB25 (RAB25, member RAS oncogene family)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR