Anti-SOX2 Mouse Monoclonal Antibody [clone: 13A220]

Lieferant: United States Biological
USBI029612EA 673 EUR
USBI029612
Anti-SOX2 Mouse Monoclonal Antibody [clone: 13A220]
Antikörper
Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: Monoclonal
Clone: 13A220
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR