Anti-SOX2 Mouse Monoclonal Antibody [clone: 13A221]
Lieferant: United States Biological
USBI029613EA 630 EUR
USBI029613
Anti-SOX2 Mouse Monoclonal Antibody [clone: 13A221]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: Monoclonal
Clone: 13A221
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse