Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 13A225]

Lieferant: United States Biological
USBI029634EA 571 EUR
USBI029634
Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 13A225]
Antikörper
Recommended dilution: Western Blot: 0.5 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: UBE2E3 (ubiquitin-conjugating enzyme E2E 3)
Clonality: Monoclonal
Clone: 13A225
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR