Anti-UCHL1 Chicken Polyclonal Antibody

Lieferant: United States Biological
USBI029638EA 527 EUR
USBI029638
Anti-UCHL1 Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.2?g/ml

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR