Anti-Chorionic Gonadotropin, Human Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029752EA 476 EUR
USBI029752
Anti-Chorionic Gonadotropin, Human Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Chorionic Gonadotropin, Human
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR