Anti-FGF20 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029777EA 411 EUR
USBI029777
Anti-FGF20 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: FGF20 (fibroblast growth factor 20)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR