Anti-GFP Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029792EA 443 EUR
USBI029792
Anti-GFP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml; Immunoprecipitation: 10-20 ?g/ml

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR