Anti-PPARGC1A Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029867EA 421 EUR
USBI029867
Anti-PPARGC1A Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: PPARGC1A (peroxisome proliferator-activated receptor gamma, coactivator 1 alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR