Anti-TGFA Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029904EA 411 EUR
USBI029904
Anti-TGFA Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Western Blot: 0.5-4μg/ml; Neutralization: 2-10 μg/ml

Type: Primary
Antigen: TGFA (transforming growth factor, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR