Anti-TNFB Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029910EA 470 EUR
USBI029910
Anti-TNFB Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Neutralization: 2-5 ?g/ml

Type: Primary
Antigen: TNFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR