Anti-ABL2 Mouse Monoclonal Antibody [clone: 13B10]
Lieferant: United States Biological
USBI029932EA 506 EUR
USBI029932
Anti-ABL2 Mouse Monoclonal Antibody [clone: 13B10]
Antikörper
Type: Primary
Antigen: ABL2 (c-abl oncogene 2, non-receptor tyrosine kinase)
Clonality: Monoclonal
Clone: 13B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human