Anti-ALCAM Mouse Monoclonal Antibody [clone: 13B26]

Lieferant: United States Biological
USBI029948EA 437 EUR
USBI029948
Anti-ALCAM Mouse Monoclonal Antibody [clone: 13B26]
Antikörper
Type: Primary
Antigen: ALCAM (activated leukocyte cell adhesion molecule)
Clonality: Monoclonal
Clone: 13B26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR