Anti-AURKA Mouse Monoclonal Antibody [clone: 13B57]

Lieferant: United States Biological
USBI029979EA 590 EUR
USBI029979
Anti-AURKA Mouse Monoclonal Antibody [clone: 13B57]
Antikörper
Type: Primary
Antigen: AURKA (aurora kinase A)
Clonality: Monoclonal
Clone: 13B57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR