Anti-BECN1 Mouse Monoclonal Antibody [clone: 13B64]

Lieferant: United States Biological
USBI029986EA 590 EUR
USBI029986
Anti-BECN1 Mouse Monoclonal Antibody [clone: 13B64]
Antikörper
Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Monoclonal
Clone: 13B64
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR