Anti-BLK Mouse Monoclonal Antibody [clone: 13B70]

Lieferant: United States Biological
USBI029992EA 380 EUR
USBI029992
Anti-BLK Mouse Monoclonal Antibody [clone: 13B70]
Antikörper
Type: Primary
Antigen: BLK (B lymphoid tyrosine kinase)
Clonality: Monoclonal
Clone: 13B70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR