Anti-Bovine Serum Albumin Mouse Monoclonal Antibody [clone: 13B81]

Lieferant: United States Biological
USBI030003EA 567 EUR
USBI030003
Anti-Bovine Serum Albumin Mouse Monoclonal Antibody [clone: 13B81]
Antikörper
Type: Primary
Antigen: Bovine Serum Albumin
Clonality: Monoclonal
Clone: 13B81
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR