Anti-CD1A Mouse Monoclonal Antibody [clone: 13B119]

Lieferant: United States Biological
USBI030041EA 437 EUR
USBI030041
Anti-CD1A Mouse Monoclonal Antibody [clone: 13B119]
Antikörper
Type: Primary
Antigen: CD1A (CD1a molecule)
Clonality: Monoclonal
Clone: 13B119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR