Anti-MS4A1 Mouse Monoclonal Antibody [clone: 13B120]

Lieferant: United States Biological
USBI030042EA 567 EUR
USBI030042
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 13B120]
Antikörper
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: Monoclonal
Clone: 13B120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR