Anti-CK5 Mouse Monoclonal Antibody [clone: 13B181]

Lieferant: United States Biological
USBI030103EA 437 EUR
USBI030103
Anti-CK5 Mouse Monoclonal Antibody [clone: 13B181]
Antikörper
Type: Primary
Antigen: CK5
Clonality: Monoclonal
Clone: 13B181
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR