Anti-CRYAB Mouse Monoclonal Antibody [clone: 13B197]

Lieferant: United States Biological
USBI030119EA 446 EUR
USBI030119
Anti-CRYAB Mouse Monoclonal Antibody [clone: 13B197]
Antikörper
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Monoclonal
Clone: 13B197
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR