Anti-KRT5 Mouse Monoclonal Antibody [clone: 13B214]

Lieferant: United States Biological
USBI030136EA 412 EUR
USBI030136
Anti-KRT5 Mouse Monoclonal Antibody [clone: 13B214]
Antikörper
Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: Monoclonal
Clone: 13B214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR