Anti-EPHB2 Mouse Monoclonal Antibody [clone: 13B256]

Lieferant: United States Biological
USBI030178EA 446 EUR
USBI030178
Anti-EPHB2 Mouse Monoclonal Antibody [clone: 13B256]
Antikörper
Type: Primary
Antigen: EPHB2 (EPH receptor B2)
Clonality: Monoclonal
Clone: 13B256
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR