Anti-MAPK6 Mouse Monoclonal Antibody [clone: 13B271]

Lieferant: United States Biological
USBI030193EA 437 EUR
USBI030193
Anti-MAPK6 Mouse Monoclonal Antibody [clone: 13B271]
Antikörper
Type: Primary
Antigen: MAPK6 (mitogen-activated protein kinase 6)
Clonality: Monoclonal
Clone: 13B271
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR