Anti-ESR1 Mouse Monoclonal Antibody [clone: 13B276]

Lieferant: United States Biological
USBI030198EA 526 EUR
USBI030198
Anti-ESR1 Mouse Monoclonal Antibody [clone: 13B276]
Antikörper
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: Monoclonal
Clone: 13B276
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR