Anti-PTK2 Mouse Monoclonal Antibody [clone: 13B290]

Lieferant: United States Biological
USBI030212EA 412 EUR
USBI030212
Anti-PTK2 Mouse Monoclonal Antibody [clone: 13B290]
Antikörper
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: Monoclonal
Clone: 13B290
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR