Anti-FRK Mouse Monoclonal Antibody [clone: 13B325]
Lieferant: United States Biological
USBI030247EA 506 EUR
USBI030247
Anti-FRK Mouse Monoclonal Antibody [clone: 13B325]
Antikörper
Type: Primary
Antigen: FRK (fyn-related kinase)
Clonality: Monoclonal
Clone: 13B325
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human