Anti-GPI Mouse Monoclonal Antibody [clone: 13B348]

Lieferant: United States Biological
USBI030270EA 446 EUR
USBI030270
Anti-GPI Mouse Monoclonal Antibody [clone: 13B348]
Antikörper
Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: Monoclonal
Clone: 13B348
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR