Anti-MKI67 Mouse Monoclonal Antibody [clone: 13B444]
Lieferant: United States Biological
USBI030366EA 526 EUR
USBI030366
Anti-MKI67 Mouse Monoclonal Antibody [clone: 13B444]
Antikörper
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: Monoclonal
Clone: 13B444
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human