Anti-KLF4 Mouse Monoclonal Antibody [clone: 13B450]

Lieferant: United States Biological
USBI030372EA 526 EUR
USBI030372
Anti-KLF4 Mouse Monoclonal Antibody [clone: 13B450]
Antikörper
Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: Monoclonal
Clone: 13B450
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR