Anti-KLHL1 Mouse Monoclonal Antibody [clone: 13B452]
Lieferant: United States Biological
USBI030374EA 526 EUR
USBI030374
Anti-KLHL1 Mouse Monoclonal Antibody [clone: 13B452]
Antikörper
Type: Primary
Antigen: KLHL1 (kelch-like family member 1)
Clonality: Monoclonal
Clone: 13B452
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human