Anti-KRT13 Mouse Monoclonal Antibody [clone: 13B461]
Lieferant: United States Biological
USBI030383EA 590 EUR
USBI030383
Anti-KRT13 Mouse Monoclonal Antibody [clone: 13B461]
Antikörper
Type: Primary
Antigen: KRT13 (keratin 13)
Clonality: Monoclonal
Clone: 13B461
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human