Anti-LIN28A Mouse Monoclonal Antibody [clone: 13B475]
Lieferant: United States Biological
USBI030397EA 567 EUR
USBI030397
Anti-LIN28A Mouse Monoclonal Antibody [clone: 13B475]
Antikörper
Type: Primary
Antigen: LIN28A (lin-28 homolog A (C. elegans))
Clonality: Monoclonal
Clone: 13B475
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human