Anti-BPIFB1 Mouse Monoclonal Antibody [clone: 13B483]
Lieferant: United States Biological
USBI030405EA 590 EUR
USBI030405
Anti-BPIFB1 Mouse Monoclonal Antibody [clone: 13B483]
Antikörper
Type: Primary
Antigen: BPIFB1 (BPI fold containing family B, member 1)
Clonality: Monoclonal
Clone: 13B483
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse