Anti-KDM1A Mouse Monoclonal Antibody [clone: 13B486]
Lieferant: United States Biological
USBI030408EA 506 EUR
USBI030408
Anti-KDM1A Mouse Monoclonal Antibody [clone: 13B486]
Antikörper
Type: Primary
Antigen: KDM1A (lysine (K)-specific demethylase 1A)
Clonality: Monoclonal
Clone: 13B486
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse