Anti-MAPK8 Mouse Monoclonal Antibody [clone: 13B506]

Lieferant: United States Biological
USBI030428EA 437 EUR
USBI030428
Anti-MAPK8 Mouse Monoclonal Antibody [clone: 13B506]
Antikörper
Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: Monoclonal
Clone: 13B506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR