Anti-MBP Mouse Monoclonal Antibody [clone: 13B511]

Lieferant: United States Biological
USBI030433EA 437 EUR
USBI030433
Anti-MBP Mouse Monoclonal Antibody [clone: 13B511]
Antikörper
Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: Monoclonal
Clone: 13B511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR